Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium iodide hydrate

10871-22 10871-14 10871-06 010871-22 010871-06 010871-14
AA10871-06EA 42.19 USD InStock
AA10871-06 CAAA10871-06 AA10871-14 CAAA10871-14 AA10871-22 CAAA10871-22
Sodium iodide dihydrate ≥99.999% (metals basis), Puratronic®
Sodium iodide dihydrate


Formula: INa·2H₂O
MW: 185.92 g/mol
Melting Pt: 64.3 °C
Density: 2.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 13517-06-1
EINECS: 231-679-3
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty