Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfate ≥99.9955% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Synonyms: Bisodium sulfateDisodium sulfateSulfuric acid disodium salt

10874-14 10874-22 010874-14 010874-22
AA10874-14EA 217.99 USD InStock
CAAA10874-14 AA10874-14 CAAA10874-22 AA10874-22
Sodium sulfate ≥99.9955% (metals basis), Puratronic®
Sodium sulphate


Formula: Na₂SO₄
MW: 142.04 g/mol
Boiling Pt: 1429 °C (1013 hPa)
Melting Pt: 884 °C
Density: 2.680
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6
EINECS: 231-820-9
Merck Index: 12,08829
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/41/16640641.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty