Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Sodium tetraborate, anhydrous ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: BoraxSodium tetraborate

010875-36 10875-22 010875-14 10875-14 10875-36 010875-22
AA10875-36EA 1081.99 USD InStock
AA10875-36 AA10875-14 CAAA10875-22 CAAA10875-14 CAAA10875-36 AA10875-22
di-Sodium tetraborate, anhydrous ≥99.998% (metals basis), Puratronic®
di-Sodium tetraborate


Formula: Na₂B₄O₇
MW: 201.22 g/mol
Boiling Pt: 1575 °C (dec.)
Melting Pt: 741 °C
Density: 2.367
Storage Temperature: Ambient
MDL Number: MFCD00081185
CAS Number: 1330-43-4
EINECS: 215-540-4
REACH: 01-2119490790-32
Merck Index: 12,08733
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty