Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Strontium fluoride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10878-06 010878-14 010878-22 10878-14 10878-22 010878-06
AA10878-06EA 58.43 USD InStock
AA10878-06 AA10878-22 CAAA10878-22 CAAA10878-06 AA10878-14 CAAA10878-14
Strontium fluoride ≥99.99% (metals basis), Puratronic®
Strontium fluoride


Formula: F₂Sr
MW: 125.62 g/mol
Boiling Pt: 2460 °C (1013 hPa)
Melting Pt: 1473 °C
Density: 4.240
Storage Temperature: Ambient
MDL Number: MFCD00011251
CAS Number: 7783-48-4
EINECS: 232-000-3
UN: 3288
ADR: 6.1,III
Merck Index: 12,09002
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty