Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid ≥33% ≥99.999999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chlorohydric acidHydrochloric acid gasHydrochlorideHydrogen chloride gasMuriatic acid

10990-AK 010990-AK 10990-AU 10990-AC 010990-AU 010990-AC
AA10990-ACEA 113.99 USD InStock
CAAA10990-AC CAAA10990-AU AA10990-AC AA10990-AU CAAA10990-AK AA10990-AK
Hydrochloric acid ≥33% ≥99.999999% (metals basis)
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 81.5…110 °C
Melting Pt: –30 °C
Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty