Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

11036.04 11036-09 11036.09 11036-04
AA11036-09EA 1316.99 USD InStock
CAAA11036-09 AA11036-09 CAAA11036-04 AA11036-04
Tetraamminepalladium(II) chloride monohydrate (≥39% Pd) ≥99.9% (metals basis)
Tetraamminepalladium(II) chloride monohydrate


Formula: Cl₂H₁₄N₄OPd
Melting Pt: 120 °C (dec.)
Density: 1,91
Storage Temperature: Ambient
MDL Number: MFCD00151033
CAS Number: 13933-31-8
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty