Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium hexachloropalladate(IV) hydrate

011039-06 11039-03 011039-03 11039-06
AA11039-03EA 216.99 USD InStock
AA11039-03 CAAA11039-06 CAAA11039-03 AA11039-06
Sodium hexachloropalladate(IV) hydrate ≥99.9% (metals basis)
Disodium hexachloropalladate hydrate


Formula: Cl₆Na₂Pd·H₂O
MW: 365.10 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00014235
CAS Number: 16010-02-9
EINECS: 258-809-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty