Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis) Pd 29%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Diammonium hexachloropalladate

011042-09 011042-04 11042-04 11042-09
AA11042-09EA 922.99 USD InStock
AA11042-09 CAAA11042-09 CAAA11042-04 AA11042-04
Ammonium hexachloropalladate(IV) (29% Pd) ≥99.9% (metals basis) Pd 29%
Ammonium hexachloropalladate(IV)


Formula: H₈Cl₆N₂Pd
Density: 2,42
Storage Temperature: Ambient
MDL Number: MFCD00015959
CAS Number: 19168-23-1
EINECS: 242-854-9
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty