Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis) Pt ≥ 51%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Ammonium tetrachloroplatinate

011046-06 011046-03 11046-03 11046-06
AA11046-03EA 160.99 USD InStock
CAAA11046-06 AA11046-03 CAAA11046-03 AA11046-06
Diammonium tetrachloroplatinate (≥51% Pt) ≥99.9% (metals basis) Pt ≥ 51%
Diammonium tetrachloroplatinate


Formula: H₈Cl₄N₂Pt
MW: 372.97 g/mol
Melting Pt: 145 °C (dec.)
Density: 2,936
Storage Temperature: Ambient
MDL Number: MFCD00010885
CAS Number: 13820-41-2
EINECS: 237-499-1
UN: 3288
ADR: 6.1,III
Merck Index: 12,00583
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty