Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Disodium hexachloroplatinate hexahydrate

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium hexachloroplatinate (IV) hexahydrate

011047-03 011047-06 11047-03 11047-06
AA11047-03EA 144.53 USD InStock
CAAA11047-06 AA11047-03 CAAA11047-03 AA11047-06
Disodium hexachloroplatinate hexahydrate
Disodium hexachloroplatinate hexahydrate
Catalyst


Formula: Cl₆Na₂Pt·6H₂O
MW: 561.87 g/mol
Melting Pt: 110 °C (-6H₂O)
Density: 2,50
Storage Temperature: Ambient
MDL Number: MFCD00149172
CAS Number: 19583-77-8
EINECS: 240-983-5
UN: 3288
ADR: 6.1,III
Merck Index: 12,08768
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty