Would you like to log in again?
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), powder -140 mesh Type 303-L
Supplier: Thermo Scientific Chemicals
Danger
011084-A1 11084-18 11084-A1 11084-30
AA11084-A1EA 270.99 USD InStock
AA11084-A1 AA11084-30 CAAA11084-A1 AA11084-18
Stainless steel (Fe:Cr:Ni; 70:17:13 wt%), powder -140 mesh Type 303-L
Stainless steel
This Stainless steel powder offers moderate corrosion resistance. Alloy 303 is frequently used in applications that require parts to be heavily machined. It is used in nuts and bolts, aircraft fittings, gears and screws.
Caution: Stable under normal conditions. Incompatible with halogens.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty