Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Barium acetate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid barium salt Barium di(acetate)

011126-14 011126-22 11126-22 11126-14
AA11126-22EA 794.99 USD InStock
AA11126-22 CAAA11126-22 AA11126-14 CAAA11126-14
Barium acetate ≥99.999% (metals basis), Puratronic®
Barium acetate


Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
Melting Pt: ∼450 °C
Density: 2.47
Storage Temperature: Ambient
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III
Merck Index: 13,00968
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty