Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Lanthanum nickel

011271-90 11271-90 011271-18 11271-18
AA11271-90EA 423.99 USD InStock
CAAA11271-90 CAAA11271-18 AA11271-90 AA11271-18
Lanthanum, compound with nickel (1:5) ≥99.9% (REO, rare earth oxide basis), REacton®
Lanthanum, compound with nickel (1:5)
Lanthanum nickel act as a superconducting intermetallic phase in lanthanum nickel boro-nitride.


Formula: LaNi₅
MW: 432.36 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00198177
CAS Number: 12196-72-4
EINECS: 235-372-5
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty