Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gadolinium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gadolinium (III) nitrate pentahydrate

11284-06 11284-22 011284-14 011284-22 011284-06 11284-14
AA11284-14EA 180.99 USD InStock
CAAA11284-14 AA11284-14 CAAA11284-06 AA11284-06 CAAA11284-22 AA11284-22
Gadolinium(III) nitrate hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Gadolinium(III) nitrate hydrate


Formula: GdN₃O₉·5H₂O
MW: 343.26 g/mol
Melting Pt: 92 °C
Density: 2.406
Storage Temperature: Ambient
MDL Number: MFCD00011023
CAS Number: 52788-53-1
EINECS: 233-437-2
UN: 1477
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty