Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid cerium (III) salt sesquihydrate Cerium (III) acetate sesquihydrate

011324-30 011324-09 11324-09 011324-18 11324-30 11324-18
AA11324-09EA 50.39 USD InStock
AA11324-09 CAAA11324-30 AA11324-30 CAAA11324-18 CAAA11324-09 AA11324-18
Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium triacetate sesquihydrate
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.


Formula: C₆H₉CeO₆₄·1,5H₂O
MW: 344.28 g/mol (317.26 anhy)
Melting Pt: 308 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051060
CAS Number: 17829-82-2
EINECS: 208-654-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty