Synonyms:
Acetic acid cerium (III) salt sesquihydrate Cerium (III) acetate sesquihydrate
011324-30
011324-09
11324-09
011324-18
11324-30
11324-18
AA11324-09EA
50.39
USD
InStock
AA11324-09
CAAA11324-30
AA11324-30
CAAA11324-18
CAAA11324-09
AA11324-18
Cerium triacetate sesquihydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Cerium triacetate sesquihydrate
Cerium(III) acetate is used in combination with bromide ion, catalyzes the liquid-phase auto-oxidation of cresols. It is used in glass manufacturing, glass Polishing, optical Glass making and polishing, pigments, pharma, Speciality chemicals electronic, oaint and driers, dyes and pigments, paper industries etc.