Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) sulfate hydrate Co ≥ 20,8%, REacton®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cobalt sulphate hydrate Cobaltous sulphate hydrate

011340-A1 011340-30 11340-A1 11340-30
AA11340-30EA 91.33 USD InStock
AA11340-30 CAAA11340-30 AA11340-A1 CAAA11340-A1
Cobalt(II) sulfate hydrate Co ≥ 20,8%, REacton®
Cobalt (II) sulphate hydrate
Ceramics, pigments, glazes, plating baths, catalyst, storage batteries, paint/ink drier


Formula: CoO₄S·H₂O
MW: 173.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149657
CAS Number: 60459-08-7
EINECS: 233-334-2
UN: 3077
ADR: 9,III
Merck Index: 12,02510
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty