Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) nitrate hexahydrate ≥97.7%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cobalt dinitrate hexahydrate Cobaltous nitrate hexahydrate

11341-A1 011341-A1 011341-14 011341-30 011341-A7 11341-A7 11341-30 11341-14
AA11341-30EA 132.8 USD InStock
CAAA11341-A1 AA11341-30 CAAA11341-30 CAAA11341-14 AA11341-A1 AA11341-A7 CAAA11341-A7 AA11341-14
Cobalt(II) nitrate hexahydrate ≥97.7%
Cobalt (II) nitrate hexahydrate


Formula: Co(NO₃)₂·6H₂O
MW: 291.03 g/mol
Melting Pt: 55…56 °C
Density: 1,87
Storage Temperature: Ambient
MDL Number: MFCD00149647
CAS Number: 10026-22-9
EINECS: 233-402-1
UN: 1477
ADR: 5.1,III
Merck Index: 13,02469
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty