Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium perrhenate (64% Re) ≥99% (metals basis) Re 64%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium perrhenate (VII)

011411-18 011411-04 11411-18 011411-09 11411-04 11411-09 11411-SS 11411-SS
AA11411-09EA 261 USD InStock
CAAA11411-SS AA11411-09 CAAA11411-09 AA11411-18 AA11411-04 CAAA11411-18 CAAA11411-04 AA11411-SS
Potassium perrhenate (64% Re) ≥99% (metals basis) Re 64%
Potassium perrhenate
A powerful oxidizer


Formula: KO₄Re
MW: 289.3 g/mol
Boiling Pt: 1360…1370 °C
Melting Pt: 550 °C
Density: 4,887
Storage Temperature: Ambient
MDL Number: MFCD00011365
CAS Number: 10466-65-6
EINECS: 233-953-8
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty