Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver phosphate ≥99%

Supplier: Thermo Scientific Chemicals
Danger

011415-14 11415-14 011415-06 11415-06
AA11415-14EA 82.8 USD InStock
AA11415-14 CAAA11415-14 AA11415-06 CAAA11415-06
Silver phosphate ≥99%
Silver phosphate
Applications: In photography
Practically insoluble in water. Slightly soluble in dilute acetic acid. Freely soluble in dilute HNO3, ammonia, ammonium carbonate, alkali cyanides and thiosulfates


Formula: Ag₃O₄P
Melting Pt: 849 °C
Density: 6,37
Storage Temperature: Ambient
MDL Number: MFCD00014148
CAS Number: 7784-09-0
EINECS: 232-049-0
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty