Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Silver (I) sulfide 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Disilver sulphide

011416-22 11416-06 11416-14 011416-06 011416-14 11416-22
AA11416-06EA 80.69 USD InStock
CAAA11416-06 AA11416-06 CAAA11416-22 AA11416-22 CAAA11416-14 AA11416-14
Silver (I) sulfide 99%
Silver (I) sulphide


Formula: Ag₂S
MW: 247.8 g/mol
Melting Pt: 825 °C
Density: 7.317
Storage Temperature: Ambient
MDL Number: MFCD00003406
CAS Number: 21548-73-2
EINECS: 244-438-2
UN: 3077
ADR: 9,III
Merck Index: 12,08677
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty