Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium fluoride trihydrate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

11496-22 11496-A3 011496-A3 011496-22 011496-36 11496-36
AA11496-22EA 48.29 USD InStock
CAAA11496-22 CAAA11496-A3 AA11496-22 CAAA11496-36 AA11496-36 AA11496-A3
Aluminium fluoride trihydrate ≥97%
Aluminium fluoride trihydrate


Formula: AlF₃·3H₂O
MW: 138.02 g/mol
Melting Pt: 250 °C (-3H₂O)
Density: 2,17
Storage Temperature: Ambient
MDL Number: MFCD00003426
CAS Number: 15098-87-0
EINECS: 232-051-1
UN: 3260
ADR: 8,III
Merck Index: 12,00351
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty