Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin(II) chloride dihydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Stannous chloride dihydrate

11536-A3 011536-36 11536-22 011536-A3 011536-22 11536-36
AA11536-36EA 176.68 USD InStock
AA11536-36 CAAA11536-22 CAAA11536-A3 AA11536-22 AA11536-A3 CAAA11536-36
Tin(II) chloride dihydrate, REacton®, Reagent Grade
Tin (II) chloride dihydrate


Formula: SnCl₂·2H₂O
MW: 225.65 g/mol
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 43…46 °C
Density: 2,71
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 600-045-1
UN: 3260
ADR: 8,III
Merck Index: 13,08861
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty