Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum disilicide Mo ≥ 61% , ∼99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Synonyms: Molybdenum silicide

11549-18 11549-30 11549.18 11549.3
AA11549-18EA 109.99 USD InStock
CAAA11549-18 AA11549-18 CAAA11549-30 AA11549-30
Molybdenum disilicide Mo ≥ 61% , ∼99.5% (metals basis)
Molybdenum disilicide
Applications: In electrical resistors, in protective coatings at high temperatures, and in engine parts in space vehicles
Insoluble in acids and aqua regia. Very soluble in HF and HNO3


Formula: MoSi₂
MW: 152.11 g/mol
Melting Pt: 1870…2030 °C
Density: 6,31
Storage Temperature: Ambient
MDL Number: MFCD00151535
CAS Number: 12136-78-6
EINECS: 235-231-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty