Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hydrogen difluoride ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium bifluoride

011557.A3 11557.36
AA11557-A3EA 215.99 USD InStock
AA11557-A3 AA11557-36
Potassium hydrogen difluoride ≥99% (metals basis)
Potassium hydrogen difluoride
Electrolyte, flux, catalyst in alkylation of benzene with olefins, and in the synthesis of pure KF


Formula: HF₂K
MW: 78.1 g/mol
Melting Pt: 239 °C (dec.)
Density: 2,370
Storage Temperature: Ambient
MDL Number: MFCD00011403
CAS Number: 7789-29-9
EINECS: 232-156-2
UN: 1811
ADR: 8,II
Merck Index: 12,07771
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty