Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc acetate dihydrate 98.0-101.0% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid zinc salt dihydrate Zinc (II) acetate dihydrate Zinc diacetate dihydrate

11559-A7 11559-A1 011559-A1 11559-A7 11559-30 011559-30
AA11559-A1EA 87.59 USD InStock
AA11559-A1 CAAA11559-A1 AA11559-30 CAAA11559-30 AA11559-A7 CAAA11559-A7
Zinc acetate dihydrate 98.0-101.0% ACS
Zinc acetate dihydrate


Formula: (H₃CCOO)₂Zn·2H₂O
MW: 219.51 g/mol
Melting Pt: 237 °C
Density: 1.735
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 14,10128
Order Now
Specification Test Results
Insoluble matter 0.005%
pH of a 5% solution 6.0-7.0 at 25°
Cl 5 ppm
SO3 0.005%
Ca 0.005%
Fe 5 ppm
Pb 0.002%
Mg 0.00005
K 0.01%
Na 0.05%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty