Synonyms:
Bisodium sulfateDisodium sulfateSulfuric acid disodium salt
011560-A9
11560-A9
011560-36
11560-36
11560-A3
011560-A3
AA11560-36EA
70.49
USD
InStock
AA11560-36
AA11560-A3
CAAA11560-A9
AA11560-A9
CAAA11560-A3
CAAA11560-36
Sodium sulfate ≥99.0%, powder ACS
Sodium sulphate
In drying organic liquids, in Kjeldahl nitrogen determination, in manufacturing of glass. Sodium sulfate anhydrous is used primarily for drying non-aqueous solvents by removing traces of water. It is used as a fining agent for molten glass, levelling agent in textile processing, and filler in detergents. It is also used in Kraft process for paper pulping, defrosting windows, manufacture of starch, glass, and soap.
Notes: Anhydrous sodium sulfate is hygroscopic in nature, and absorbs moisture converting to the hydrated form. The hydrated form on heating gets converted back to anhydrous form.