Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tetrafluoroborate ≥95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium fluoroborate

11562-A3 011562-A3 011562-36 11562-36
AA11562-A3EA 163.99 USD InStock
AA11562-A3 CAAA11562-36 CAAA11562-A3 AA11562-36
Sodium tetrafluoroborate ≥95%
Sodium tetrafluoroborate
Fluorinating agent


Formula: BF₄Na
MW: 109.79 g/mol
Melting Pt: 384 °C (dec.)
Density: 2,47
Storage Temperature: Ambient
MDL Number: MFCD00003515
CAS Number: 13755-29-8
EINECS: 237-340-6
Merck Index: 12,08761
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty