Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chromium(III) nitrate nonahydrate ≥98.5%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Chromic nitrate nonahydrate Chromium trinitrate nonahydrate

11565-14 11565-A1 11565-30 011565-A1 011565-14 011565-30
AA11565-30EA 79.68 USD InStock
CAAA11565-A1 AA11565-30 CAAA11565-30 CAAA11565-14 AA11565-A1 AA11565-14
Chromium(III) nitrate nonahydrate ≥98.5%
Chromium (III) nitrate nonahydrate
Catalyst, corrosion inhibitor, in textile printing


Formula: CrN₃O₉·9H₂O
MW: 400.15 g/mol
Melting Pt: 60 °C
Density: 1,807 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149659
CAS Number: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1,III
Merck Index: 13,02246
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty