Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc phosphate hydrate, tech.

Supplier: Thermo Scientific Chemicals

011589.A3 11589-36 11589.36 11589-A3
AA11589-A3EA 104.99 USD InStock
CAAA11589-A3 AA11589-A3 CAAA11589-36 AA11589-36
Zinc phosphate hydrate, tech.
Zinc phosphate hydrate
Soluble in acetic acid, dilute mineral acids, ammonia. Insoluble in water


Formula: O₈P₂Zn₃.xH₂O
MW: 386.08 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00036282
CAS Number: 7779-90-0
EINECS: 231-944-3
UN: 3077
ADR: 9,III
Merck Index: 14,10151
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty