Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

di-Potassium hydrogen phosphate ≥98.0% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Dipotassium hydrogen phosphatePotassium hydrogen phosphate

11593-30 011593-30 11593-A7 11593-A1 011593-A7 011593-A1
AA11593-A1EA 251.26 USD InStock
CAAA11593-A1 AA11593-A1 CAAA11593-30 AA11593-30 AA11593-A7 CAAA11593-A7
di-Potassium hydrogen phosphate ≥98.0% ACS
di-Potassium hydrogen phosphate


Formula: K₂HPO₄
MW: 174.18 g/mol
Melting Pt: 340 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011383
CAS Number: 7758-11-4
EINECS: 231-834-5
Merck Index: 13,07743
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/42/16640642.jpg
Order Now
Specification Test Results
Insoluble matter 0.01%
Loss on drying at 105°C 1%
pH of a 5% solution 8.5-9.6 at 25°
Cl 0.003%
Nitrogen compounds (as N) 0.001%
SO3 0.005%
Heavy Metals (as Pb) 5 ppm max.
Fe 0.001%
Na 0.05%%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty