Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium acetate ≥97.0% ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Acetic acid ammonium salt

11599-36 11599-A3 011599-A3 011599-36
AA11599-36EA 107.52 USD InStock
AA11599-36 AA11599-A3 CAAA11599-36 CAAA11599-A3
Ammonium acetate ≥97.0% ACS
Ammonium acetate
Reagent in analytical chemistry, drugs, textile dying, preserving meats, foam rubbers, vinyl plastics


Formula: H₃CCOONH₄
MW: 77.08 g/mol
Melting Pt: ∼114 °C (dec.)
Density: 1.17
Flash Pt: 136 °C (277 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8
EINECS: 211-162-9
Merck Index: 13,00495
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/04/16640604.jpg
Order Now
Specification Test Results
Insoluble matter 0.0005%
Residue after ignition 0.01%
pH of a 5% solution 6.7-7.3 at 25°
Cl 5 ppm
NO3 0.001%
SO3 0.001%
Heavy Metals (as Pb) 5 ppm max.
Fe 5 ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty