Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper(I) iodide 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cuprous iodide Copper iodide

011606-A1 011606-30 11606-A1 11606-30
AA11606-A1EA 332.99 USD InStock
AA11606-A1 AA11606-30 CAAA11606-A1 CAAA11606-30
Copper(I) iodide 98%
Copper (I) iodide
As a catalyst in organic synthesis, cathode ray tube coating, temperature indicator with HgI2


Formula: CuI
MW: 190.45 g/mol
Boiling Pt: ∼1290 °C
Melting Pt: 605 °C
Density: 5.63
Storage Temperature: Ambient
MDL Number: MFCD00010978
CAS Number: 7681-65-4
EINECS: 231-674-6
UN: 3077
ADR: 9,III
Merck Index: 13,02692
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty