Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nickel(II) perchlorate hexahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Nickel diperchlorate hexahydrate Perchloric acid nickel (II) salt hexahydrate

011631-36 11631-22 011631-22 11631-14 011631-14 11631-36
AA11631-36EA 162.99 USD InStock
AA11631-36 AA11631-14 CAAA11631-14 CAAA11631-36 AA11631-22 CAAA11631-22
Nickel(II) perchlorate hexahydrate, REacton®, Reagent Grade
Nickel (II) perchlorate hexahydrate


Formula: Cl₂NiO₈·6H₂O
MW: 365.68 g/mol
Melting Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00149804
CAS Number: 13520-61-1
EINECS: 237-124-1
UN: 1481
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty