Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) perchlorate hexahydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Cobalt diperchlorate hexahydrate

11652-18 11652-09 011652-30 011652-18 11652-30 011652-09
AA11652-18EA 76.49 USD InStock
CAAA11652-18 CAAA11652-09 AA11652-18 AA11652-09 CAAA11652-30 AA11652-30
Cobalt(II) perchlorate hexahydrate, REacton®, Reagent Grade
Cobalt (II) perchlorate hexahydrate


Formula: Cl₂CoO₈·6H₂O
MW: 365.93 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00150207
CAS Number: 13478-33-6
EINECS: 236-653-5
UN: 1481
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty