Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium perchlorate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

011658-A1 011658-30
AA11658-30EA 75.69 USD InStock
AA11658-30 AA11658-A1
Ammonium perchlorate, REacton®, Reagent Grade
Ammonium perchlorate


Formula: H₄ClNO₄
MW: 117.49 g/mol
Melting Pt: Dec.
Density: 1.95
Storage Temperature: Ambient
MDL Number: MFCD00011421
CAS Number: 7790-98-9
EINECS: 232-235-1
UN: 1442
ADR: 5.1,II
Merck Index: 12,00574
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty