Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium sulfide hydrate, tech.

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium sulphide hydrate

11664-36 011664-A3 11664-A3 011664-36
AA11664-36EA 76.12 USD InStock
AA11664-36 AA11664-A3 CAAA11664-36 CAAA11664-A3
Sodium sulfide hydrate, tech.
Sodium sulphide hydrate
Sodium sulfide hydrate is used as an oxygen scavenger in water treatment, bleaching agent in textile industry, desulfurising and as a dechlorinating agent and in leather industry, sulfonation and sulfomethylation agent in chemical industry.

It is involved in the production of rubber chemicals and sulfur dyes. It is also used to prepare 3-nitroaniline from 1,3-dinitrobenzene and thioethers from alkyl halide. It plays an important role in other applications such as ore flotation, oil recovery, making dyes, and detergent. In addition, it is used in pulp and paper industry in the Kraft process.


Formula: Na₂S·1H₂O
MW: 96.06 g/mol (anhy)
Boiling Pt: ∼920 °C (1013 hPa)
Melting Pt: ∼ 50 °C (dec.)
Density: 1.427
Storage Temperature: Ambient
MDL Number: MFCD00149183
CAS Number: 27610-45-3
EINECS: 215-211-5
UN: 1849
ADR: 8,II
REACH: 01-2119513694-38
Merck Index: 12,08829
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty