Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ruthenium(III) chloride oxide (≥35,0% Ru), ammoniated

Supplier: Thermo Scientific Chemicals
Synonyms: Ruthenium RedRuthenium oxychlorideTetraamminechlorohydroxoruthenium(III) chloride

11806-06 11806-03 11806.06 11806.03
AA11806-06EA 499.99 USD InStock
AA11806-06 CAAA11806-03 CAAA11806-06 AA11806-03
Ruthenium(III) chloride oxide (≥35,0% Ru), ammoniated
Ruthenium(III) chloride oxide
Applications: Microscopic stain
Soluble in water, ammonia


Formula: H₄₂Cl₆N₁₄O₂Ru₃
MW: 786.354 g/mol
Melting Pt: >500 °C (dec.)
Density: 3,110
Storage Temperature: Ambient
MDL Number: MFCD00011479
CAS Number: 11103-72-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty