Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphotungstic acid hydrate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Dodecawolframophosphoric acid hydrate Wolframophosphoric acid hydrate Phosphotungstic acid hydrate 12-Tungstophosphate hydrate

011827-30 011827-18 11827-18 11827-30
AA11827-30EA 394.5 USD InStock
CAAA11827-30 AA11827-30 AA11827-18 CAAA11827-18
Phosphotungstic acid hydrate, REacton®, Reagent Grade
Phosphotungstic acid hydrate


Formula: H₃[P(W₃O₁₀)₄]·nH₂O
MW: 2898.07 g/mol
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00011342
CAS Number: 12501-23-4
EINECS: 215-682-7
UN: 3260
ADR: 8,III
Merck Index: 12,07521
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty