Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium hexacyanoruthenate(II) hydrate (≥23.0% Ru)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium hexacyanoruthenate hydrate

11876-04 011876-09 011876-SS 011876-04 11876-SS 11876-09
AA11876-SSEA 52.39 USD InStock
CAAA11876-SS AA11876-SS CAAA11876-04 AA11876-04 CAAA11876-09 AA11876-09
Potassium hexacyanoruthenate(II) hydrate (≥23.0% Ru)
Potassium hexacyanoruthenate(II) hydrate


Formula: C₆H₂K₄N₆ORu
MW: 413.57 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00168064
CAS Number: 339268-21-2
EINECS: 239-097-1
UN: 1588
ADR: 6.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty