Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium perruthenate(VII) ≥97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium perruthenate

11877-03 011877-06 11877-06 011877-03
AA11877-03EA 191.99 USD InStock
CAAA11877-03 AA11877-03 CAAA11877-06 AA11877-06
Potassium perruthenate(VII) ≥97%
Potassium perruthenate(VII)


Formula: C₃₂H₃₆N₂O₅
MW: 528.649 g/mol
Melting Pt: 440 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00061333
CAS Number: 10378-50-4
EINECS: 233-835-6
UN: 1479
ADR: 5.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty