Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dipotassium hexachloropalladate Pd ≥ 26,3%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Potassium hexachloropalladate(IV)

011884-06 11884-06 11884-03 011884-03
AA11884-03EA 83.59 USD InStock
CAAA11884-06 AA11884-03 AA11884-06 CAAA11884-03
Dipotassium hexachloropalladate Pd ≥ 26,3%
Dipotassium hexachloropalladate


Formula: Cl₆K₂Pd
MW: 397.33 g/mol
Melting Pt: Dec.
Density: 2,738
Storage Temperature: Ambient
MDL Number: MFCD00043081
CAS Number: 16919-73-6
EINECS: 240-974-6
UN: 3077
ADR: 9,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty