Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc ≥99.994% (metals basis), foil, Puratronic®, Thickness 0.1 mm (0.004 in)

Supplier: Thermo Scientific Chemicals

11912-GH 11912-FI 11912-HG 011912-FI 011912-GH 011912-HG
AA11912-FIEA 99.39 USD InStock
AA11912-FI CAAA11912-FI AA11912-GH CAAA11912-GH AA11912-HG CAAA11912-HG
Zinc ≥99.994% (metals basis), foil, Puratronic®, Thickness 0.1 mm (0.004 in)
Zinc


Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908 °C (1013 hPa)
Melting Pt: 420 °C
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty