Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palladium ≥99.95% (metals basis), powder , -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

012068-06 12068-03 012068-03 012068-14 12068-06 12068-14
AA12068-06EA 1518.18 USD InStock
AA12068-06 CAAA12068-06 AA12068-03 AA12068-14 CAAA12068-03 CAAA12068-14
Palladium ≥99.95% (metals basis), powder , -200 mesh
Palladium


Formula: Pd
MW: 106.420 g/mol
Density: 12.02 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3
EINECS: 231-115-6
UN: 3089
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty