Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zirconium diboride ≥99.5% (metals basis excluding Hf)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Zirconium boride

12140-18 12140.3 12140.18 12140-30
AA12140-30EA 482.99 USD InStock
CAAA12140-18 AA12140-30 CAAA12140-30 AA12140-18
Zirconium diboride ≥99.5% (metals basis excluding Hf)
Zirconium diboride


Formula: B₂Zr
Melting Pt: 3000 °C
Storage Temperature: Ambient
MDL Number: MFCD00064648
CAS Number: 12045-64-6
EINECS: 234-963-5
UN: 3178
ADR: 4.1,III
Order Now
Specification Test Results
Form -325 Mesh Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty