Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Potassium tetrachloroaurate hydrate ≥99.99% (metals basis) Au ≥ 49%, Premion®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gold potassium chloride hydratePotassium tetrachloroaurate(III) hydratePotassium gold(III) chloride hydrate

12150-06 012150-03 12150-03 012150-06
AA12150-03EA 144.99 USD InStock
AA12150-03 CAAA12150-06 CAAA12150-03 AA12150-06
Potassium tetrachloroaurate hydrate ≥99.99% (metals basis) Au ≥ 49%, Premion®
Potassium tetrachloroaurate hydrate
Soluble in water


Formula: AuCl₄K·H₂O
MW: 377.88 g/mol (anhy)
Melting Pt: 357 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043088
CAS Number: 27988-75-6
EINECS: 237-190-1
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty