Synonyms:
Gold(iii)hydroxide
12159-03
12159-06
12159-09
012159-09
012159-03
012159-06
AA12159-03EA
219.99
USD
InStock
CAAA12159-03
CAAA12159-06
AA12159-03
AA12159-06
CAAA12159-09
AA12159-09
Gold trihydroxide Au ≥ 79% (≥79% Au)
Gold trihydroxide
Applications: Gold plating solutions, porcelain decoration
Decomposed by light to metallic gold
Gold plating solutions, porcelain decoration