Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99.95% (metals basis), Premion®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium platinum chloride hydrateSodium chloroplatinate(II) hydrateDisodium tetrachloroplatinate hydrate

012168-06 12168-03 12168-06 012168-03
AA12168-06EA 712.99 USD InStock
AA12168-06 CAAA12168-06 AA12168-03 CAAA12168-03
Sodium tetrachloroplatinate(II) hydrate (≥42,4% Pt) ≥99.95% (metals basis), Premion®
Sodium tetrachloroplatinate(II) hydrate


Formula: Cl₄H₂Na₂OPt
MW: 382.88 g/mol (anhy)
Melting Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150188
CAS Number: 207683-21-4
EINECS: 233-051-4
Order Now
Specification Test Results
Form Crystalline

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty