Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Sodium chloroosmate(IV) dihydrateDisodium hexachloroosmate dihydrate

012176-06 012176-03 012176-01 12176-06 12176-03 12176-01
AA12176-06EA 672.99 USD InStock
AA12176-06 AA12176-03 CAAA12176-06 CAAA12176-01 AA12176-01 CAAA12176-03
Sodium hexachloroosmate(IV) dihydrate Os ≥38,7%
Disodium hexachloroosmate


Formula: Cl₆Na₂Os
MW: 484.93 g/mol (448.90 anhy)
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 389…393 °C
Density: 3,59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00014234
CAS Number: 1307-81-9
EINECS: 215-152-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty