Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum carbide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals

012192-22 12192-36 12192-22 012192-36
AA12192-36EA 454.99 USD InStock
AA12192-36 AA12192-22 CAAA12192-36 CAAA12192-22
Molybdenum carbide ≥99.5% (metals basis)
Molybdenum carbide


Formula: CMo₂
Melting Pt: 2687 °C
Storage Temperature: Ambient
MDL Number: MFCD00014218
CAS Number: 12069-89-5
EINECS: 235-115-7
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty